Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010188.5 | NP_034318.2 | 5 | 753 | ||
GenBank mRNA | AF197930.1 | - | 5 | 748 | ||
AK077227.1 | - | 5 | 713 | |||
AK151557.1 | - | 5 | 713 | |||
AK151795.1 | - | 5 | 713 | |||
AK152875.1 | - | 5 | 713 | |||
AK153384.1 | - | 5 | 713 | |||
AK153449.1 | - | 5 | 713 | |||
AK154892.1 | - | 5 | 714 | |||
AK159919.1 | - | 5 | 699 | |||
AK163401.1 | - | 3 | 703 | |||
AK169887.1 | - | 5 | 710 | |||
AK171289.1 | - | 5 | 714 | |||
AY897419.1 | - | 5 | 702 | |||
AY897420.1 | - | 5 | 698 | |||
AY897421.1 | - | 5 | 702 | |||
AY897422.1 | - | 5 | 702 | |||
AY897423.1 | - | 5 | 702 | |||
AY897424.1 | - | 5 | 697 | |||
AY897425.1 | - | 5 | 707 | |||
AY897426.1 | - | 5 | 707 | |||
AY897427.1 | - | 5 | 707 | |||
AY897428.1 | - | 5 | 711 | |||
AY897429.1 | - | 5 | 702 | |||
BC052819.1 | - | 5 | 707 | 30044058 | ||
M14215.1 | - | 5 | 748 |