Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dopey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177208.3 | NP_796182.2 | 21 | 5332 | ||
GenBank mRNA | BC039975.1 | - | 1 | 697 |