Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153081.3 | NP_694721.2 | 4, 5 | 1486 | ||
GenBank mRNA | AB041591.1 | - | 4 | 1364 | ||
BC130255.1 | - | 3, 4 | 989 |