Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110512.1 | NP_001103982.1 | 9 | 825 | ||
NM_145524.3 | NP_663499.2 | 10 | 1289 | |||
GenBank mRNA | AK087927.1 | - | 9 | 1169 | ||
BC004636.1 | - | 11 | 1336 | 3585719 | ||
BC057960.1 | - | 10 | 1156 | 5354784 | ||
DQ093357.1 | - | 8 | 1150 | |||
DQ093358.1 | - | 10 | 1265 | |||
DQ230973.1 | - | 9 | 1057 | |||
DQ230974.1 | - | 9 | 829 |