Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122640.1 | NP_001116112.1 | 9 | 793 | ||
NM_001122641.1 | NP_001116113.1 | 9 | 793 | |||
NM_001122642.1 | NP_001116114.1 | 9 | 793 | |||
NM_001122643.1 | NP_001116115.1 | 9 | 793 | |||
NM_144529.2 | NP_653112.2 | 9 | 793 | |||
GenBank mRNA | AB060553.1 | - | 9 | 791 | ||
AB060554.1 | - | 9 | 791 | |||
AK036468.1 | - | 9 | 730 | |||
AK036617.1 | - | 9 | 730 | |||
AK147010.1 | - | 9 | 764 | |||
AK149986.1 | - | 9 | 765 | |||
AK152266.1 | - | 9 | 730 | |||
AK152301.1 | - | 9 | 757 | |||
AK161636.1 | - | 9 | 794 | |||
AK195035.1 | - | 1 | 44 | |||
BC003259.1 | - | 9 | 741 | 3500822 |