Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099918.1 | NP_001093388.1 | 6 | 580 | ||
GenBank mRNA | AK015258.1 | - | 2 | 134 | ||
AK019564.1 | - | 2 | 106 | |||
DQ113419.1 | - | 6 | 580 | |||
HQ713436.1 | - | 6 | 580 | |||
JF429832.1 | - | 6 | 593 |