Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114662.1 | NP_001108134.1 | 9 | 1567 | ||
NM_172510.4 | NP_766098.2 | 9 | 1567 | |||
GenBank mRNA | AK038882.1 | - | 9 | 1460 | ||
AK044667.1 | - | 9 | 1441 | |||
AK048692.1 | - | 8 | 1369 | |||
AK085484.1 | - | 6 | 983 | |||
AK162495.1 | - | 9 | 1471 | |||
BC058790.1 | - | 9 | 1416 | 6395070 | ||
FN434126.1 | - | 2 | 100 |