Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114662.1 | NP_001108134.1 | 6 | 1268 | ||
NM_172510.4 | NP_766098.2 | 6 | 1268 | |||
GenBank mRNA | AK038882.1 | - | 6 | 1161 | ||
AK044667.1 | - | 6 | 1142 | |||
AK048692.1 | - | 5 | 1070 | |||
AK162495.1 | - | 6 | 1172 | |||
BC058790.1 | - | 6 | 1117 | 6395070 |