Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321729.1 | NP_001308658.1 | 7 | 919 | ||
NM_001321730.1 | NP_001308659.1 | 7 | 919 | |||
NM_001321731.1 | NP_001308660.1 | 7 | 919 | |||
NM_001321733.1 | NP_001308662.1 | 7 | 919 | |||
NM_001321734.1 | NP_001308663.1 | 6 | 753 | |||
NM_015189.2 | NP_056004.1 | 7 | 919 | |||
NR_135773.1 | 7 | 919 | ||||
NR_135774.1 | 7 | 919 | ||||
XM_011532711.2 | XP_011531013.1 | 7 | 915 | |||
XM_011532712.2 | XP_011531014.1 | 7 | 915 | |||
XM_017003641.1 | XP_016859130.1 | 3 | 353 | |||
GenBank mRNA | AB023136.1 | - | 7 | 902 | ||
AK026474.1 | - | 4 | 414 |