Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085546.2 | NP_001079015.1 | 3, 4 | 267 | ||
GenBank mRNA | CF161713.1 | - | 3, 4 | 267 | ||
CF164689.1 | - | 3, 4 | 261 |