Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kansl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081045.1 | NP_001074514.1 | 2 | 1398 | ||
GenBank mRNA | AK037800.1 | - | 3 | 964 | ||
AK122484.1 | - | 2 | 1404 | |||
AK153679.1 | - | 2 | 1379 | |||
AK161514.1 | - | 2 | 913 | |||
BC043121.1 | - | 2 | 947 | 6409084 | ||
BC053389.1 | - | 2 | 948 | 30061629 | ||
BC054752.1 | - | 3 | 1221 | 6834541 | ||
BC079594.1 | - | 2 | 946 | 6827379 |