Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 14 | 1924 | ||
NM_018752.3 | NP_061222.3 | 14 | 1817 | |||
GenBank mRNA | AF047714.1 | - | 14 | 1686 | ||
AK044507.1 | - | 14 | 1848 | |||
AK148193.1 | - | 12 | 1766 | |||
AK148268.1 | - | 14 | 1671 | |||
AK148275.1 | - | 14 | 1727 | |||
AK149290.1 | - | 14 | 1846 | |||
AK192969.1 | - | 1 | 50 | |||
AK193962.1 | - | 1 | 50 | |||
AK194126.1 | - | 4 | 245 | |||
AK196486.1 | - | 4 | 245 | |||
AK198455.1 | - | 1 | 50 | |||
AK199086.1 | - | 1 | 50 | |||
AK200460.1 | - | 1 | 50 | |||
AK202089.1 | - | 4 | 250 | |||
AK203960.1 | - | 4 | 245 | |||
AK204100.1 | - | 4 | 245 | |||
AK205038.1 | - | 1 | 50 | |||
AK217026.1 | - | 1 | 50 | |||
AY180104.2 | - | 14 | 1924 |