Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 15 | 1710 | ||
GenBank mRNA | AK051425.1 | - | 15 | 1711 | ||
AK141083.1 | - | 9 | 730 | |||
AK143518.1 | - | 15 | 1738 | |||
AK145483.1 | - | 15 | 1740 | |||
AK166188.1 | - | 15 | 1919 | |||
BC011069.1 | - | 15 | 1453 | 4006745 | ||
BC013663.1 | - | 15 | 1769 | 4208974 | ||
X69025.1 | - | 15 | 1732 |