Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201529.2 | NP_963287.2 | 17 | 3468 | ||
GenBank mRNA | AK129231.1 | - | 14 | 3253 | ||
AK137694.1 | - | 8 | 1707 | |||
BC049164.1 | - | 11 | 2053 | |||
BC082553.1 | - | 15 | 3483 | |||
BC145011.1 | - | 10 | 1871 | 9053559 | ||
EF071857.1 | - | 17 | 3439 |