Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201529.2 | NP_963287.2 | 22, 23 | 3947 | ||
GenBank mRNA | AK129231.1 | - | 19, 20 | 3732 | ||
AK137694.1 | - | 13, 14 | 2186 | |||
BC082553.1 | - | 20, 21 | 3962 | |||
BC145011.1 | - | 15, 16 | 2431 | 9053559 | ||
EF071857.1 | - | 22, 23 | 3918 |