Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081009.1 | NP_001074478.1 | 10 | 770 | ||
GenBank mRNA | AK149889.1 | - | 9 | 737 | ||
AK149918.1 | - | 9 | 737 | |||
AK150113.1 | - | 9 | 683 | |||
AK150208.1 | - | 9 | 737 | |||
AK150732.1 | - | 9 | 615 | |||
AK151510.1 | - | 9 | 615 | |||
AK152919.1 | - | 9 | 615 | |||
AK153169.1 | - | 9 | 615 | |||
AK153270.1 | - | 9 | 615 | |||
BC021881.1 | - | 9 | 740 | 3710450 | ||
EF591877.1 | - | 503 |