Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111111.1 | NP_001104581.1 | 17, 18 | 1853 | ||
GenBank mRNA | AB476647.1 | - | 16, 17 | 1729 | ||
AB476648.1 | - | 17, 18 | 1792 | |||
AK010773.1 | - | 16, 17 | 1774 | |||
AK131109.1 | - | 1787 | ||||
AK155125.1 | - | 17, 18 | 1843 | |||
AK157410.1 | - | 17, 18 | 1833 | |||
AK158186.1 | - | 16, 17 | 1962 | |||
BC060054.1 | - | 16, 17 | 2349 |