Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175664.3 | NP_783595.1 | 1 | 102 | ||
GenBank mRNA | BC116853.1 | - | 1 | 102 | 40126172 | |
BC116857.1 | - | 1 | 102 | 40126176 |