Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033123.3 | NP_001028295.2 | 2, 3 | 214 | ||
GenBank mRNA | BC086907.1 | - | 2, 3 | 214 |
Gene Symbol: 6230416C02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018090.1 | - | 2, 3 | 273 | ||
BC056163.1 | - | 2, 3 | 208 |
Gene Symbol: Etohi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177399.1 | NP_001170870.1 | 2, 3 | 232 | ||
NM_001177400.1 | NP_001170871.1 | 2, 3 | 232 | |||
GenBank mRNA | AK005093.1 | - | 2, 3 | 233 |
Gene Symbol: Gm14327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099593.1 | - | 2, 3 | 292 |
Gene Symbol: Gm14420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177568.1 | NP_001171039.1 | 2, 3 | 241 | ||
GenBank mRNA | BC124559.1 | - | 2, 3 | 241 | 30730885 |
Gene Symbol: Gm14431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177406.1 | NP_001170877.1 | 2, 3 | 246 |
Gene Symbol: Gm14436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242943.1 | NP_001229872.1 | 197 |
Gene Symbol: Gm14440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199308.1 | NP_001186237.1 | 2, 3 | 213 |
Gene Symbol: Gm8898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177407.1 | NP_001170878.1 | 2, 3 | 246 |
Gene Symbol: Gm8923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242944.1 | NP_001229873.1 | 2, 3 | 232 |