Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290497.1 | NP_001277426.1 | 4 | 635 | ||
NM_001290498.1 | NP_001277427.1 | 4 | 613 | |||
NM_177102.4 | NP_796076.1 | 4 | 783 | |||
GenBank mRNA | AK079312.1 | - | 4 | 784 | ||
BC058996.1 | - | 4 | 732 | 5711063 |