Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026767.4 | NP_081043.1 | 2 | 399 | ||
GenBank mRNA | AI527477.1 | - | 1 | 343 | ||
AK003223.1 | - | 2 | 309 | |||
BC037761.1 | - | 1 | 343 | |||
CJ237809.1 | - | 2 | 399 |