Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025250.3 | NP_001020421.2 | 4 | 1348 | ||
NM_001025257.3 | NP_001020428.2 | 4 | 1348 | |||
NM_001110266.1 | NP_001103736.1 | 1 | 658 | |||
NM_001110267.1 | NP_001103737.1 | 1 | 658 | |||
NM_001110268.1 | NP_001103738.1 | 1 | 658 | |||
NM_001287056.1 | NP_001273985.1 | 4 | 1348 | |||
NM_001287057.1 | NP_001273986.1 | 4 | 1348 | |||
NM_001287058.1 | NP_001273987.1 | 4 | 1348 | |||
NM_009505.4 | NP_033531.3 | 4 | 1348 | |||
GenBank mRNA | AY120866.1 | - | 2 | 226 | ||
AY707864.1 | - | 5 | 545 | |||
AY750956.1 | - | 5 | 506 | |||
AY750957.1 | - | 5 | 469 | |||
AY756068.1 | - | 4 | 284 | |||
BC061468.1 | - | 4 | 721 | 6816435 | ||
BQ554097.1 | - | 1 | 303 | |||
BQ832724.1 | - | 3 | 249 | |||
EF028705.1 | - | 5 | 541 | |||
M95200.1 | - | 4 | 413 | |||
S37052.1 | - | 4 | 339 | |||
S38083.1 | - | 4 | 339 | |||
S38100.1 | - | 4 | 339 |