Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025250.3 | NP_001020421.2 | 8 | 1728 | ||
NM_001025257.3 | NP_001020428.2 | 6 | 1524 | |||
NM_001110266.1 | NP_001103736.1 | 5 | 1038 | |||
NM_001110267.1 | NP_001103737.1 | 4 | 966 | |||
NM_001110268.1 | NP_001103738.1 | 3 | 834 | |||
NM_001287056.1 | NP_001273985.1 | 8 | 1728 | |||
NM_001287057.1 | NP_001273986.1 | 7 | 1656 | |||
NM_001287058.1 | NP_001273987.1 | 6 | 1524 | |||
NM_009505.4 | NP_033531.3 | 7 | 1656 | |||
GenBank mRNA | AW913188.1 | - | 3 | 286 | ||
BC061468.1 | - | 7 | 1029 | 6816435 | ||
BQ832724.1 | - | 5 | 425 | |||
M95200.1 | - | 7 | 721 |