Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122662.1 | NP_001116134.1 | 2 | 1420 | ||
GenBank mRNA | AK163212.1 | - | 2 | 1420 |
Gene Symbol: Gm5662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013824.3 | NP_001013846.1 | 2 | 1417 | ||
GenBank mRNA | AK146341.1 | - | 2 | 1420 |