Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015889.2 | NP_001015889.1 | 2 | 192 | ||
NM_027139.5 | NP_081415.1 | 2 | 184 | |||
GenBank mRNA | AF305839.1 | - | 1 | 82 | ||
BC042723.1 | - | 2 | 159 | 5006430 | ||
BC043028.1 | - | 1 | 16 | 5692221 | ||
BC096683.1 | - | 2 | 182 | |||
BU946135.1 | - | 2 | 211 | |||
BY108476.1 | - | 2 | 184 |
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027592.3 | NP_081868.1 | 2 | 184 | ||
GenBank mRNA | AK014903.1 | - | 1 | 106 | ||
BC019453.1 | - | 2 | 135 | 4024616 |