Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026195.3 | NP_080471.2 | 14 | 1571 | ||
GenBank mRNA | AK010611.2 | - | 14 | 1554 | ||
AK012074.1 | - | 14 | 1508 | |||
AK076091.1 | - | 8 | 863 | |||
BC039925.1 | - | 14 | 1577 | 3155580 |