Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030595.1 | NP_085098.1 | 9, 10 | 1881 | ||
GenBank mRNA | Y18276.1 | - | 9, 10 | 1881 |