Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025574.1 | NP_001020745.1 | 5 | 1054 | ||
GenBank mRNA | AK017304.1 | - | 5 | 1072 | ||
BC083121.1 | - | 5 | 1054 | 6314132 |
Gene Symbol: Bpifb9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175167.3 | NP_780376.2 | 5 | 1046 | ||
GenBank mRNA | AK029377.1 | - | 5 | 1072 | ||
BC047134.1 | - | 5 | 1046 | 6308426 | ||
BC080814.1 | - | 5 | 1055 | 6391119 |