Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183320.3 | NP_899143.2 | 5 | 1539 | ||
GenBank mRNA | AK029697.1 | - | 1540 | |||
BC115710.1 | - | 1534 | 40058794 |
Gene Symbol: AV320801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177918.1 | NP_808586.1 | 1544 | |||
GenBank mRNA | AK029939.1 | - | 1545 |
Gene Symbol: Gm15023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099321.2 | NP_001092791.1 | 5 | 1526 | ||
GenBank mRNA | BC115763.1 | - | 5 | 1526 | 40058793 | |
BC119584.1 | - | 5 | 1526 | 40058853 | ||
BC119585.1 | - | 5 | 1526 | 40058855 |
Gene Symbol: Gm7903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242937.1 | NP_001229866.1 | 1539 |
Gene Symbol: Pramel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031390.2 | NP_113567.1 | 5 | 1543 | ||
GenBank mRNA | AY004873.1 | - | 5 | 1557 |