Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098267.1 | NP_001091737.1 | 2 | 1354 | ||
GenBank mRNA | AM690745.1 | - | 2 | 1354 |