Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033351.2 | NP_001028523.1 | 9 | 3875 | ||
NM_001276355.1 | NP_001263284.1 | 9 | 3935 | |||
GenBank mRNA | AK173314.1 | - | 9 | 2936 | ||
BQ180214.1 | - | 3 | 460 |