Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016671.3 | NP_057880.2 | 8 | 1065 | ||
GenBank mRNA | AK040457.1 | - | 6 | 1161 | ||
AK133753.1 | - | 8 | 1080 |