Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098723.1 | NP_001092193.1 | 1 | 1041 | ||
GenBank mRNA | AK161451.1 | - | 1 | 1043 |
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036071.1 | - | 6 | 1893 |