Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098227.1 | NP_001091697.1 | 6 | 586 | ||
NM_016807.2 | NP_058087.2 | 6 | 583 | |||
GenBank mRNA | AF003693.1 | - | 6 | 578 | ||
AF077527.1 | - | 6 | 542 | |||
AK014678.1 | - | 560 | ||||
AK028429.1 | - | 6 | 559 | |||
AK033412.1 | - | 6 | 584 | |||
AK146085.1 | - | 6 | 558 | |||
AK150278.1 | - | 6 | 551 | |||
AK150321.1 | - | 6 | 554 | |||
AK150544.1 | - | 6 | 558 | |||
AK150623.1 | - | 6 | 562 | |||
AK150661.1 | - | 6 | 551 | |||
AK150707.1 | - | 6 | 557 | |||
AK150721.1 | - | 6 | 566 | |||
AK150736.1 | - | 6 | 551 | |||
AK150801.1 | - | 6 | 556 | |||
AK150996.1 | - | 6 | 558 | |||
AK151050.1 | - | 6 | 558 | |||
AK152001.1 | - | 6 | 557 | |||
AK152017.1 | - | 6 | 558 | |||
AK152080.1 | - | 6 | 565 | |||
AK152434.1 | - | 6 | 558 | |||
AK152552.1 | - | 6 | 562 | |||
AK152577.1 | - | 6 | 557 | |||
AK152855.1 | - | 6 | 558 | |||
AK153042.1 | - | 6 | 558 | |||
AK159124.1 | - | 6 | 554 | |||
AK159538.1 | - | 6 | 556 | |||
AK159677.1 | - | 6 | 565 | |||
AK165657.1 | - | 6 | 553 | |||
AK167513.1 | - | 6 | 554 | |||
AK168004.1 | - | 6 | 561 | |||
AK168777.1 | - | 6 | 554 | |||
AK169219.1 | - | 6 | 563 | |||
AK169415.1 | - | 6 | 592 | |||
AK169687.1 | - | 6 | 558 | |||
AK171162.1 | - | 6 | 558 | |||
AK178827.1 | - | 2 | 144 | |||
BC019400.1 | - | 6 | 549 | 4017442 |