Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 13 | 2062 | ||
NM_001128607.1 | NP_001122079.1 | 13 | 1972 | |||
NM_183428.3 | NP_906273.3 | 16 | 3486 | |||
GenBank mRNA | AB370261.1 | - | 13 | 1904 | ||
AK133437.1 | - | 3 | 257 | |||
AK147266.1 | - | 8 | 1015 | |||
AK147644.1 | - | 8 | 1015 | |||
AK220462.1 | - | 13 | 1969 | |||
BC043337.1 | - | 2 | 109 | |||
BC068138.1 | - | 13 | 1932 | |||
BC079875.1 | - | 13 | 1891 | 6844082 | ||
CB526050.1 | - | 6 | 429 | |||
L00919.1 | - | 17 | 3561 | |||
LK021332.1 | - | 12 | 1280 | |||
LK021333.1 | - | 12 | 1280 |