Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 6, 7 | 1116 | ||
NM_001128607.1 | NP_001122079.1 | 6, 7 | 1026 | |||
NM_183428.3 | NP_906273.3 | 6, 7 | 2378 | |||
GenBank mRNA | AB370261.1 | - | 5, 6 | 901 | ||
AK147266.1 | - | 1, 2 | 72 | |||
AK147644.1 | - | 1, 2 | 72 | |||
AK220462.1 | - | 6, 7 | 1023 | |||
BC068138.1 | - | 6, 7 | 986 | |||
BC079875.1 | - | 6, 7 | 945 | 6844082 | ||
CN537562.1 | - | 4, 5 | 434 | |||
L00919.1 | - | 7, 8 | 2453 | |||
LK021332.1 | - | 4, 5 | 271 | |||
LK021333.1 | - | 4, 5 | 271 |