Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 9, 10 | 1592 | ||
NM_001128607.1 | NP_001122079.1 | 9, 10 | 1502 | |||
NM_183428.3 | NP_906273.3 | 9, 10 | 2854 | |||
GenBank mRNA | AB370261.1 | - | 8, 9 | 1377 | ||
AK147266.1 | - | 4, 5 | 548 | |||
AK147644.1 | - | 4, 5 | 548 | |||
AK220462.1 | - | 9, 10 | 1499 | |||
BC068138.1 | - | 9, 10 | 1462 | |||
BC079875.1 | - | 9, 10 | 1421 | 6844082 | ||
CN526385.1 | - | 3, 4 | 436 | |||
L00919.1 | - | 10, 11 | 2929 | |||
LK021332.1 | - | 7, 8 | 747 | |||
LK021333.1 | - | 7, 8 | 747 |