Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177243.4 | NP_796217.2 | 9 | 1097 | ||
GenBank mRNA | AK078966.1 | - | 9 | 1098 | ||
AY034145.1 | - | 8 | 1010 |