Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164727.1 | NP_001158199.1 | 6 | 677 | ||
GenBank mRNA | BC172015.1 | - | 5 | 568 | 9054301 | |
EF651799.1 | - | 3 | 308 | |||
EF651804.1 | - | 4 | 473 | |||
EF651806.1 | - | 4 | 473 | |||
EF651807.1 | - | 4 | 473 | |||
EF651833.1 | - | 308 |
Gene Symbol: 2610042L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011748.1 | - | 5 | 515 | ||
BC147228.1 | - | 4 | 424 | 9056284 | ||
EF651843.1 | - | 2 | 179 |
Gene Symbol: Gm2897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177715.2 | NP_001171186.1 | 6 | 712 | ||
GenBank mRNA | BC028273.1 | - | 6 | 722 |
Gene Symbol: Gm3194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF651800.1 | - | 3 | 308 |
Gene Symbol: Gm3383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291093.1 | NP_001278022.1 | 5 | 569 | ||
NM_001291094.1 | NP_001278023.1 | 381 | ||||
GenBank mRNA | EF651835.1 | - | 5 | 569 |
Gene Symbol: Gm3500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256886.1 | NP_001243815.1 | 6 | 702 | ||
GenBank mRNA | EF651805.1 | - | 473 |
Gene Symbol: Gm3696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024712.2 | NP_001019883.2 | 677 | |||
GenBank mRNA | BC055874.1 | - | 676 | 3491523 | ||
EF651802.1 | - | 308 | ||||
EF651822.1 | - | 3 | 308 | |||
EF651829.1 | - | 308 |
Gene Symbol: Gm3739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF651817.1 | - | 3 | 308 |
Gene Symbol: Gm5796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029930.2 | NP_001025101.2 | 535 | |||
GenBank mRNA | BC093494.1 | - | 535 | 2812218 |
Gene Symbol: LOC100036572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027416.1 | - | 1472 |
Gene Symbol: LOC100861615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270812.1 | NP_001257741.1 | 5 | 528 | ||
GenBank mRNA | AK049619.1 | - | 5 | 529 |