Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015201.4 | NP_056016.1 | 16 | 2353 | ||
GenBank mRNA | AB060694.1 | - | 15 | 2021 | ||
AK024840.1 | - | 12 | 1448 | |||
AK290677.1 | - | 16 | 2315 | |||
AK293719.1 | - | 15 | 1997 | |||
AK310015.1 | - | 14 | 1927 | |||
BC001086.2 | - | 15 | 2079 | |||
BC005160.2 | - | 15 | 2106 | |||
BC007274.1 | - | 16 | 2248 | |||
BC013787.1 | - | 16 | 2315 | 3872212 | ||
BC013980.2 | - | 16 | 2318 | 3633864 | ||
BC017674.2 | - | 16 | 2305 | 3905068 | ||
BC073813.1 | - | 11 | 1612 | |||
BC098446.1 | - | 12 | 1749 | |||
D50914.1 | - | 15 | 2093 |