Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297673.1 | NP_001284602.1 | 8 | 1908 | ||
NM_001297674.1 | NP_001284603.1 | 8 | 1908 | |||
NM_015203.4 | NP_056018.2 | 9 | 1986 | |||
XM_005245033.2 | XP_005245090.1 | 9 | 1949 | |||
XM_006711240.2 | XP_006711303.1 | 9 | 1949 | |||
XM_011509344.2 | XP_011507646.1 | 9 | 1949 | |||
XM_017000763.1 | XP_016856252.1 | 8 | 1871 | |||
XM_017000764.1 | XP_016856253.1 | 9 | 1324 | |||
XM_017000765.1 | XP_016856254.1 | 9 | 1949 | |||
XM_017000766.1 | XP_016856255.1 | 8 | 1583 | |||
XM_017000767.1 | XP_016856256.1 | 8 | 1871 | |||
GenBank mRNA | AB007929.2 | - | 9 | 1355 | ||
AK056707.1 | - | 9 | 1393 | |||
AK125864.1 | - | 9 | 1556 | |||
AK291703.1 | - | 9 | 1404 | |||
AK304380.1 | - | 8 | 1326 | |||
AK308148.1 | - | 8 | 1317 | |||
BC045623.1 | - | 8 | 1309 | 5269891 | ||
BX641025.1 | - | 10 | 1881 |