Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145073.2 | NP_659539.1 | 1 | 340 | ||
GenBank mRNA | BC125355.1 | - | 1 | 336 | 40129870 | |
BC125357.1 | - | 1 | 336 | 40129872 |
Gene Symbol: Hist1h3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178204.2 | NP_835511.1 | 1 | 351 | ||
GenBank mRNA | AK020421.1 | - | 352 | |||
Z30939.1 | - | 1 | 78 |
Gene Symbol: Hist1h3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178205.2 | NP_835512.1 | 1 | 312 | ||
GenBank mRNA | BC127042.1 | - | 1 | 305 |
Gene Symbol: Hist1h3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178206.2 | NP_835513.1 | 1 | 343 | ||
GenBank mRNA | BC107285.2 | - | 1 | 310 |
Gene Symbol: Hist1h3i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178207.2 | NP_835514.1 | 1 | 339 | ||
GenBank mRNA | AK155722.1 | - | 340 | |||
BC107287.2 | - | 1 | 308 | |||
BC116383.2 | - | 1 | 312 | 40101629 | ||
BC127148.1 | - | 1 | 311 |