Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037247.4 | NP_001032324.1 | 2 | 388 | ||
GenBank mRNA | AK005872.1 | - | 2 | 343 | ||
AY591385.1 | - | 2 | 252 | |||
DQ012042.1 | - | 2 | 388 |