Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AY761185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012640.2 | NP_001012658.1 | 1 | 193 | ||
GenBank mRNA | AY761185.1 | - | 1 | 155 | ||
BC141303.1 | - | 1 | 193 | 9055915 |