Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp54c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100110.1 | NP_001093580.1 | 557 | |||
GenBank mRNA | AK146826.1 | - | 595 | |||
BC005543.1 | - | 574 | 3585216 |
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011899.4 | NP_036029.2 | 5, 6 | 631 | ||
GenBank mRNA | AK011928.1 | - | 5, 6 | 595 | ||
BC019683.1 | - | 5, 6 | 560 | 3708250 |
Gene Symbol: Srp54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100109.1 | NP_001093579.1 | 5, 6 | 867 | ||
GenBank mRNA | AK136033.1 | - | 5, 6 | 868 | ||
X16319.1 | - | 596 |