Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081171.2 | NP_001074640.1 | 73 | 10231 | ||
GenBank mRNA | AK129157.1 | - | 34 | 5005 | ||
AK158204.1 | - | 6 | 858 | |||
BC020313.2 | - | 10 | 1403 | |||
BC043478.1 | - | 3 | 377 | |||
BC062207.1 | - | 11 | 1664 | |||
CN702366.1 | - | 2 | 220 | |||
U37501.1 | - | 73 | 9925 |