Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134674.1 | NP_001128146.1 | 6 | 671 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163011.1 | NP_001156483.1 | 6 | 656 | ||
GenBank mRNA | BC093515.1 | - | 1056 |
Gene Symbol: Mup7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134675.1 | NP_001128147.1 | 6 | 720 |