Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081315.1 | NP_001074784.1 | 2, 3 | 1705 | ||
GenBank mRNA | AK147555.1 | - | 2, 3 | 1704 | ||
BC157915.1 | - | 1, 2 | 1470 | 9088158 | ||
BC157957.1 | - | 1, 2 | 1470 | |||
BC172121.1 | - | 1, 2 | 1470 |