Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133800.3 | NP_598561.2 | 2 | 222 | ||
GenBank mRNA | AK036891.1 | - | 2 | 165 | ||
AK037035.1 | - | 2 | 216 | |||
AK083539.1 | - | 224 | ||||
AK167446.1 | - | 2 | 216 | |||
AY940114.1 | - | 2 | 165 | |||
BC013701.1 | - | 2 | 207 | 3483364 | ||
BC094424.1 | - | 2 | 209 | 6819926 |