Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015210.3 | NP_056025.2 | 3 | 249 | ||
XM_005258098.3 | XP_005258155.1 | 2 | 1272 | |||
XM_005258099.4 | XP_005258156.1 | 2 | 1272 | |||
XM_011525640.2 | XP_011523942.1 | 2 | 1272 | |||
XM_011525641.2 | XP_011523943.1 | 2 | 1272 | |||
XM_017025671.1 | XP_016881160.1 | 2 | 641 | |||
XM_017025672.1 | XP_016881161.1 | 2 | 242 | |||
XM_017025673.1 | XP_016881162.1 | 3 | 249 | |||
XM_017025674.1 | XP_016881163.1 | 2 | 296 | |||
XM_017025675.1 | XP_016881164.1 | 3 | 249 | |||
GenBank mRNA | AB018345.2 | - | 2 | 289 | ||
AK131528.1 | - | 3 | 249 |